Assay Details
Target Gene Details
Entrez Gene ID: | 100533182 |
Gene Name: | SEC24B antisense RNA 1 |
Gene Aliases: |
1/2-SBSRNA4 |
Location: |
Chr.4:109429963-109433817 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC24B-AS1 | NR_039978.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv595129 | Chr.4:109000908 - 109672304 on Build GRCh38 | Gain |
|
| nsv4461 | Chr.4:109401813 - 109435744 on Build GRCh38 | Insertion |
|

Genomic Map