Assay Details
Target Gene Details
Entrez Gene ID: | 51574 |
Gene Name: | La ribonucleoprotein domain family member 7 |
Gene Aliases: |
ALAZS, HDCMA18P, PIP7S |
Location: |
Chr.4:112636964-112657592 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARP7 | NM_001267039.1 | NP_001253968.1 | ||
| NM_015454.2 | NP_056269.1 | |||
| NM_016648.3 | NP_057732.2 | |||
| NR_049768.1 | ||||
| AK000274.1 | ||||
| BC006981.1 | AAH06981.1 | |||
| BC066945.1 | AAH66945.1 | |||
| BX647100.1 | ||||
| CN346208.1 | ||||
| DB270203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006030 | Chr.4:112618215 - 112736658 on Build GRCh38 | Gain |
|
| nsv525715 | Chr.4:112615590 - 112657453 on Build GRCh38 | Loss |
|
| nsv1014926 | Chr.4:112574087 - 112770975 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map