Assay Details
Target Gene Details
Entrez Gene ID: | 166929 |
Gene Name: | sphingomyelin synthase 2 |
Gene Aliases: |
SMS2 |
Location: |
Chr.4:107823645-107915047 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGMS2 | XM_011531701.2 | 1 | 614 | XP_011530003.1 |
| XM_011531702.2 | 1 | 614 | XP_011530004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830031 | Chr.4:107724456 - 107883976 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map