Assay Details
Target Gene Details
Entrez Gene ID: | 9353 |
Gene Name: | slit guidance ligand 2 |
Gene Aliases: |
SLIL3, Slit-2 |
Location: |
Chr.4:20251905-20620561 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLIT2 | NM_001289135.2 | 1 | 348 | NP_001276064.1 |
| NM_001289136.2 | 1 | 348 | NP_001276065.1 | |
| NM_004787.3 | 1 | 348 | NP_004778.1 | |
| XM_005248211.3 | 1 | 348 | XP_005248268.1 | |
| XM_006713986.3 | 1 | 348 | XP_006714049.1 | |
| DA721496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950554 | Chr.4:20250778 - 20255977 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map