Assay Details
Target Gene Details
Entrez Gene ID: | 6164 |
Gene Name: | ribosomal protein L34 |
Gene Aliases: |
L34 |
Location: |
Chr.4:108620558-108630484 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL34 | NM_000995.4 | NP_000986.2 | ||
| NM_001319232.1 | NP_001306161.1 | |||
| NM_001319234.1 | NP_001306163.1 | |||
| NM_001319235.1 | NP_001306164.1 | |||
| NM_001319236.1 | NP_001306165.1 | |||
| NM_033625.3 | NP_296374.1 | |||
| AA128015.1 | ||||
| AA128058.1 | ||||
| AF147334.1 | ||||
| AY762981.1 | AAV88043.1 | |||
| BC001773.1 | AAH01773.1 | |||
| BC070208.1 | AAH70208.1 | |||
| BC106009.1 | AAI06010.1 | |||
| BE394007.1 | ||||
| BM971961.1 | ||||
| BU588998.1 | ||||
| CB132938.1 | ||||
| CD175155.1 | ||||
| CD249923.1 | ||||
| CR542232.1 | CAG47028.1 | |||
| CR542242.1 | CAG47038.1 | |||
| DA779124.1 | ||||
| L38941.1 | AAC41916.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs202169763] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map