Assay Details
Target Gene Details
Entrez Gene ID: | 55132 |
Gene Name: | La ribonucleoprotein domain family member 1B |
Gene Aliases: |
LARP2 |
Location: |
Chr.4:128060655-128222931 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5385n100 | Chr.4:127882004 - 128173863 on Build GRCh38 | Gain |
|
| nsv595388 | Chr.4:127932758 - 128163676 on Build GRCh38 | Gain |
|
| dgv9259n54 | Chr.4:128060930 - 128091483 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map