Assay Details
Target Gene Details
Entrez Gene ID: | 55435 |
Gene Name: | adaptor related protein complex 1 associated regulatory protein |
Gene Aliases: |
2C18, C4orf16, GBAR, PRO0971, gamma-BAR |
Location: |
Chr.4:112231727-112273110 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP1AR | NM_001128426.2 | NP_001121898.1 | ||
| NM_018569.5 | NP_061039.3 | |||
| NR_136622.1 | ||||
| XM_017008378.1 | XP_016863867.1 | |||
| XM_017008379.1 | XP_016863868.1 | |||
| XM_017008380.1 | XP_016863869.1 | |||
| AK310787.1 | ||||
| AK315299.1 | ||||
| AL136628.1 | CAB66563.1 | |||
| BC009485.2 | AAH09485.1 | |||
| BM823928.1 | ||||
| BX647702.1 | CAH56145.1 | |||
| DA463845.1 |
More Information
Set Membership: |
|

Genomic Map