Assay Details
Target Gene Details
Entrez Gene ID: | 23324 |
Gene Name: | mannosidase alpha class 2B member 2 |
Gene Aliases: |
- |
Location: |
Chr.4:6575174-6622403 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAN2B2 | NM_001292038.1 | NP_001278967.1 | ||
| NM_015274.2 | NP_056089.1 | |||
| AB023152.2 | BAA76779.2 | |||
| AK025803.1 | ||||
| AK301473.1 | ||||
| AL832232.1 | ||||
| AL833071.2 | CAD89971.1 | |||
| BC033307.1 | AAH33307.1 | |||
| BC094773.1 | AAH94773.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829849 | Chr.4:6503821 - 6698228 on Build GRCh38 | Loss |
|
| nsv949778 | Chr.4:6560274 - 6626273 on Build GRCh38 | Deletion |
|
| nsv470006 | Chr.4:6576760 - 6615221 on Build GRCh38 | Loss |
|
| nsv1007724 | Chr.4:6586380 - 6611657 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map