Assay Details
Target Gene Details
Entrez Gene ID: | 6164 |
Gene Name: | ribosomal protein L34 |
Gene Aliases: |
L34 |
Location: |
Chr.4:108620558-108630484 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL34 | NM_000995.4 | NP_000986.2 | ||
| NM_033625.3 | NP_296374.1 | |||
| BC001773.1 | AAH01773.1 | |||
| BM971961.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3601724 | Chr.4:108629616 - 108652513 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map