Assay Details
Target Gene Details
Entrez Gene ID: | 57721 |
Gene Name: | methyltransferase like 14 |
Gene Aliases: |
- |
Location: |
Chr.4:118685370-118712270 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL14 | NM_020961.3 | NP_066012.1 | ||
| XM_005263166.4 | XP_005263223.1 | |||
| XM_011532162.2 | XP_011530464.1 | |||
| AB046847.1 | ||||
| AK055555.1 | BAB70954.1 | |||
| AK296056.1 | ||||
| BC006565.2 | AAH06565.1 | |||
| BC007449.2 | AAH07449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759279 | Chr.4:117557693 - 118753587 on Build GRCh38 | Gain |
|
| nsv519017 | Chr.4:117907652 - 118850326 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map