Assay Details
Target Gene Details
Entrez Gene ID: | 92689 |
Gene Name: | family with sequence similarity 114 member A1 |
Gene Aliases: |
Noxp20 |
Location: |
Chr.4:38867733-38945744 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM114A1 | NM_138389.2 | NP_612398.2 | ||
| NR_033290.1 | ||||
| XM_005262672.2 | XP_005262729.1 | |||
| XM_006714033.2 | XP_006714096.1 | |||
| XM_017008838.1 | XP_016864327.1 | |||
| AK292831.1 | ||||
| AK296859.1 | ||||
| AK304050.1 | ||||
| BC040452.1 | AAH40452.1 | |||
| DA802033.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73236654] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map