Assay Details
Target Gene Details
Entrez Gene ID: | 101930370 |
Gene Name: | uncharacterized LOC101930370 |
Gene Aliases: |
- |
Location: |
Chr.4:173131933-173169652 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101930370 | NR_134241.1 | |||
| NR_134242.1 |

Genomic Map