Assay Details
Target Gene Details
Entrez Gene ID: | 10393 |
Gene Name: | anaphase promoting complex subunit 10 |
Gene Aliases: |
APC10, DOC1 |
Location: |
Chr.4:144967117-145098541 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANAPC10 | NM_001256706.1 | 5 | 1344 | NP_001243635.1 |
| NM_001256707.1 | 6 | 1417 | NP_001243636.1 | |
| NM_001256708.1 | 5 | 1550 | NP_001243637.1 | |
| NM_001256709.1 | 7 | 1485 | NP_001243638.1 | |
| NM_001256710.1 | 6 | 1382 | NP_001243639.1 | |
| NM_001256711.1 | 7 | 1455 | NP_001243640.1 | |
| NM_001256712.1 | 4 | 1212 | NP_001243641.1 | |
| NM_001318367.1 | 5 | 1382 | NP_001305296.1 | |
| NM_014885.4 | 5 | 1333 | NP_055700.2 | |
| NR_046345.1 | 3 | 1096 | ||
| XM_011531524.2 | 6 | 1573 | XP_011529826.1 | |
| XM_011531529.2 | XP_011529831.1 | |||
| XM_011531531.2 | 4 | 1239 | XP_011529833.1 | |
| XM_011531532.2 | XP_011529834.1 | |||
| XM_017007638.1 | 5 | 1360 | XP_016863127.1 | |
| XM_017007639.1 | 6 | 2009 | XP_016863128.1 | |
| XM_017007640.1 | 5 | 1401 | XP_016863129.1 | |
| XM_017007643.1 | 6 | 1398 | XP_016863132.1 | |
| XM_017007644.1 | 6 | 1372 | XP_016863133.1 | |
| AW025455.1 | 1 | 125 |
More Information
Set Membership: |
|

Genomic Map