Assay Details
Target Gene Details
Entrez Gene ID: | 1909 |
Gene Name: | endothelin receptor type A |
Gene Aliases: |
ET-A, ETA, ETA-R, ETAR, ETRA, MFDA, hET-AR |
Location: |
Chr.4:147480917-147544954 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EDNRA | NM_001166055.1 | NP_001159527.1 | ||
| NM_001256283.1 | NP_001243212.1 | |||
| NM_001957.3 | NP_001948.1 | |||
| NR_045958.1 | ||||
| AK304451.1 | ||||
| AK312812.1 | ||||
| AK315931.1 | ||||
| BC022511.1 | AAH22511.1 | |||
| D90348.1 | BAA14359.1 | |||
| DA956937.1 | ||||
| S45956.1 | AAB23644.1 | |||
| S57498.1 | AAB25530.2 | |||
| X61950.1 | CAA43953.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830109 | Chr.4:147373909 - 147547453 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map