Assay Details
Target Gene Details
Entrez Gene ID: | 6059 |
Gene Name: | ATP binding cassette subfamily E member 1 |
Gene Aliases: |
ABC38, OABP, RLI, RLI1, RNASEL1, RNASELI, RNS4I |
Location: |
Chr.4:145098004-145129524 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCE1 | NM_001040876.1 | NP_001035809.1 | ||
| NM_002940.2 | NP_002931.2 | |||
| AB062293.1 | BAB93476.1 | |||
| AK315198.1 | ||||
| BC016283.1 | AAH16283.1 | |||
| BT009779.1 | AAP88781.1 | |||
| X74987.1 | CAA52920.1 | |||
| X76388.1 | CAA53972.1 |
More Information
Set Membership: |
|

Genomic Map