Assay Details
Target Gene Details
Entrez Gene ID: | 3512 |
Gene Name: | joining chain of multimeric IgA and IgM |
Gene Aliases: |
IGCJ, IGJ, JCH |
Location: |
Chr.4:70655541-70676901 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JCHAIN | NM_144646.3 | NP_653247.1 | ||
| XM_006714211.2 | XP_006714274.1 | |||
| XM_006714212.2 | XP_006714275.1 | |||
| XM_011531925.1 | XP_011530227.1 | |||
| XM_011531926.1 | XP_011530228.1 | |||
| AK312014.1 | ||||
| BC038982.1 | AAH38982.1 | |||
| BP261870.1 | ||||
| DQ884395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829965 | Chr.4:70630001 - 70806591 on Build GRCh38 | Gain |
|
| nsv1012495 | Chr.4:69979953 - 70947088 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map