Assay Details
Target Gene Details
Entrez Gene ID: | 25854 |
Gene Name: | family with sequence similarity 149 member A |
Gene Aliases: |
MST119, MSTP119 |
Location: |
Chr.4:186104798-186172663 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM149A | XM_017007996.1 | XP_016863485.1 | ||
| XM_017007997.1 | XP_016863486.1 | |||
| AK057166.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894090 | Chr.4:186011626 - 186212421 on Build GRCh38 | Gain |
|
| nsv470097 | Chr.4:186010756 - 186210350 on Build GRCh38 | Gain |
|
| nsv1031258 | Chr.4:186058545 - 186139101 on Build GRCh38 | Gain |
|
| nsv1028146 | Chr.4:185582745 - 186417181 on Build GRCh38 | Gain |
|
| nsv830172 | Chr.4:186091611 - 186247270 on Build GRCh38 | Loss |
|
| dgv683n27 | Chr.4:186008072 - 186210350 on Build GRCh38 | Gain |
|
| nsv461864 | Chr.4:186085652 - 186335850 on Build GRCh38 | Gain |
|
| esv3603517 | Chr.4:186008703 - 186213065 on Build GRCh38 | Gain |
|
| dgv1052e214 | Chr.4:186072002 - 186212619 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72708987] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map