Assay Details
Target Gene Details
Entrez Gene ID: | 80216 |
Gene Name: | alpha kinase 1 |
Gene Aliases: |
8430410J10Rik, LAK |
Location: |
Chr.4:112286420-112442618 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALPK1 | XM_017008638.1 | XP_016864127.1 | ||
| XM_017008641.1 | XP_016864130.1 |
More Information
Set Membership: |
|

Genomic Map