Assay Details
Target Gene Details
Entrez Gene ID: | 399 |
Gene Name: | ras homolog family member H |
Gene Aliases: |
ARHH, TTF |
Location: |
Chr.4:40191011-40244764 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOH | NM_001278364.1 | NP_001265293.1 | ||
| NM_001278365.1 | NP_001265294.1 | |||
| NM_001278366.1 | NP_001265295.1 | |||
| NM_001278367.1 | NP_001265296.1 | |||
| NM_001278368.1 | NP_001265297.1 | |||
| NM_001278369.1 | NP_001265298.1 | |||
| AW402442.1 | ||||
| AY673954.1 | ||||
| BI818548.1 | ||||
| BM457874.1 | ||||
| DA951252.1 | ||||
| DB115332.1 |
More Information
Set Membership: |
|

Genomic Map