Assay Details
Target Gene Details
Entrez Gene ID: | 2160 |
Gene Name: | coagulation factor XI |
Gene Aliases: |
FXI |
Location: |
Chr.4:186265945-186290727 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F11 | NM_000128.3 | NP_000119.1 | ||
| XM_005262821.3 | XP_005262878.1 | |||
| XM_005262822.3 | XP_005262879.1 | |||
| XM_005262823.3 | XP_005262880.1 | |||
| XM_006714137.2 | XP_006714200.1 | |||
| XM_017007884.1 | XP_016863373.1 | |||
| XM_017007885.1 | XP_016863374.1 | |||
| XM_017007886.1 | XP_016863375.1 | |||
| AF045649.1 | AAC24506.1 | |||
| AK313195.1 | ||||
| BC119014.1 | ||||
| BC122863.1 | ||||
| M13142.1 | AAA52487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034638 | Chr.4:186264231 - 186292323 on Build GRCh38 | Loss |
|
| nsv1028146 | Chr.4:185582745 - 186417181 on Build GRCh38 | Gain |
|
| nsv461864 | Chr.4:186085652 - 186335850 on Build GRCh38 | Gain |
|
| nsv596401 | Chr.4:186151229 - 186445495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map