Assay Details
Target Gene Details
Entrez Gene ID: | 55153 |
Gene Name: | SDA1 domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.4:75949906-75990962 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDAD1 | NM_001288983.1 | NP_001275912.1 | ||
| NM_001288984.1 | NP_001275913.1 | |||
| NM_018115.3 | NP_060585.2 | |||
| XM_005263102.4 | XP_005263159.1 | |||
| XM_005263105.4 | XP_005263162.1 | |||
| XM_017008354.1 | XP_016863843.1 | |||
| XM_017008355.1 | XP_016863844.1 | |||
| AK001360.1 | BAA91648.1 | |||
| AK022683.1 | BAB14177.1 | |||
| AK300074.1 | ||||
| AK307540.1 | ||||
| BC054040.1 | AAH54040.1 | |||
| BC063797.1 | AAH63797.1 | |||
| CR749574.1 | CAH18368.1 | |||
| DN602051.1 |
More Information
Set Membership: |
|

Genomic Map