Assay Details
Target Gene Details
Entrez Gene ID: | 441009 |
Gene Name: | CPEB2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.4:14909961-15002045 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPEB2-AS1 | NR_038857.1 |

Genomic Map