Assay Details
Target Gene Details
Entrez Gene ID: | 57728 |
Gene Name: | WD repeat domain 19 |
Gene Aliases: |
ATD5, CED4, DYF-2, IFT144, NPHP13, ORF26, Oseg6, PWDMP, SRTD5 |
Location: |
Chr.4:39182404-39285810 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR19 | NM_001317924.1 | NP_001304853.1 | ||
| NM_025132.3 | NP_079408.3 | |||
| XM_011513724.1 | XP_011512026.1 | |||
| XM_011513725.2 | XP_011512027.1 | |||
| XM_011513726.2 | XP_011512028.1 | |||
| XM_011513727.1 | XP_011512029.1 | |||
| XM_017008501.1 | XP_016863990.1 | |||
| AK026780.1 | BAB15550.1 | |||
| AK225588.1 | ||||
| AK294730.1 | ||||
| AK304161.1 | ||||
| AY029257.1 | AAK38745.1 | |||
| BC032578.1 | AAH32578.1 |
More Information
Set Membership: |
|

Genomic Map