Assay Details
Target Gene Details
Entrez Gene ID: | 101929595 |
Gene Name: | uncharacterized LOC101929595 |
Gene Aliases: |
- |
Location: |
Chr.4:107863479-107932119 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929595 | NR_125929.1 | |||
| AK123292.1 |
Target Gene Details
Entrez Gene ID: | 166929 |
Gene Name: | sphingomyelin synthase 2 |
Gene Aliases: |
SMS2 |
Location: |
Chr.4:107823645-107915047 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGMS2 | NM_001136258.1 | NP_001129730.1 | ||
| XM_011531698.2 | XP_011530000.1 | |||
| XM_011531699.2 | XP_011530001.1 | |||
| XM_011531700.2 | XP_011530002.1 | |||
| XM_011531701.2 | XP_011530003.1 | |||
| XM_011531702.2 | XP_011530004.1 | |||
| XM_017007839.1 | XP_016863328.1 | |||
| XM_017007840.1 | XP_016863329.1 | |||
| XM_017007841.1 | XP_016863330.1 | |||
| AF452717.1 | AAP13352.1 | |||
| AK290344.1 | ||||
| DC417367.1 |
More Information
Set Membership: |
|

Genomic Map