Assay Details
Target Gene Details
Entrez Gene ID: | 128 |
Gene Name: | alcohol dehydrogenase 5 (class III), chi polypeptide |
Gene Aliases: |
ADH-3, ADHX, FALDH, FDH, GSH-FDH, GSNOR, HEL-S-60p |
Location: |
Chr.4:99070978-99088788 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADH5 | NM_000671.4 | NP_000662.3 | ||
| AK129570.1 | ||||
| AK312367.1 | ||||
| BC070491.1 | ||||
| BQ881561.1 | ||||
| DA737788.1 | ||||
| M29872.1 | AAA51597.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002927 | Chr.4:99031321 - 99227533 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map