Assay Details
Target Gene Details
Entrez Gene ID: | 126 |
Gene Name: | alcohol dehydrogenase 1C (class I), gamma polypeptide |
Gene Aliases: |
ADH3 |
Location: |
Chr.4:99336492-99353045 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADH1C | NM_000669.4 | 1 | 202 | NP_000660.1 |
| NR_133005.1 | 1 | 202 | ||
| XM_011531588.2 | 1 | 94 | XP_011529890.1 | |
| XM_011531589.1 | 1 | 202 | XP_011529891.1 | |
| DA562068.1 | 1 | 202 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5345n100 | Chr.4:99181544 - 99523288 on Build GRCh38 | Gain |
|
| nsv528518 | Chr.4:99336998 - 99353592 on Build GRCh38 | Loss |
|
| nsv1002809 | Chr.4:99312891 - 99364351 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map