Assay Details
Target Gene Details
Entrez Gene ID: | 10983 |
Gene Name: | cyclin I |
Gene Aliases: |
CCNI1, CYC1, CYI |
Location: |
Chr.4:77047155-77076013 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNI | NM_006835.2 | NP_006826.1 | ||
| XM_011531555.1 | XP_011529857.1 | |||
| AB209766.1 | BAD93003.1 | |||
| AF135162.1 | AAF43786.1 | |||
| AK301111.1 | ||||
| AK312632.1 | ||||
| BC000420.2 | AAH00420.1 | |||
| BC004975.1 | AAH04975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1073775 | Chr.4:77074746 - 77076047 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map