Assay Details
Target Gene Details
Entrez Gene ID: | 53407 |
Gene Name: | syntaxin 18 |
Gene Aliases: |
Ufe1 |
Location: |
Chr.4:4418968-4542338 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STX18 | NM_016930.2 | NP_058626.1 | ||
| XM_011513480.1 | XP_011511782.1 | |||
| XM_011513483.1 | XP_011511785.1 | |||
| XM_011513485.2 | XP_011511787.1 | |||
| XM_017008297.1 | XP_016863786.1 | |||
| AB028741.1 | BAA95213.1 | |||
| AK310026.1 | ||||
| AL832867.1 | ||||
| AY453396.1 | AAS47513.1 | |||
| BC014613.2 | AAH14613.1 | |||
| BT007150.1 | AAP35814.1 | |||
| BT020135.1 | AAV38937.1 | |||
| BX647083.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73793287] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map