Assay Details
Target Gene Details
Entrez Gene ID: | 10983 |
Gene Name: | cyclin I |
Gene Aliases: |
CCNI1, CYC1, CYI |
Location: |
Chr.4:77047155-77076013 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNI | XM_011531555.1 | 5 | 2039 | XP_011529857.1 |
More Information
Set Membership: |
|

Genomic Map