Assay Details
Target Gene Details
Entrez Gene ID: | 27065 |
Gene Name: | neuron specific gene family member 1 |
Gene Aliases: |
D4S234, D4S234E, NEEP21, P21 |
Location: |
Chr.4:4380988-4419058 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSG1 | XM_017008022.1 | XP_016863511.1 | ||
| AK094573.1 | ||||
| BC034227.1 | AAH34227.1 |
More Information
Set Membership: |
|

Genomic Map