Assay Details
Target Gene Details
Entrez Gene ID: | 166614 |
Gene Name: | doublecortin like kinase 2 |
Gene Aliases: |
CL2, CLICK-II, CLICK2, CLIK2, DCAMKL2, DCDC3, DCDC3B, DCK2 |
Location: |
Chr.4:150078274-150258563 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCLK2 | NM_001040260.3 | NP_001035350.2 | ||
| NM_001040261.4 | NP_001035351.4 | |||
| NR_036614.1 | ||||
| XM_005262785.2 | XP_005262842.1 | |||
| XM_005262786.2 | XP_005262843.1 | |||
| XM_017007832.1 | XP_016863321.1 | |||
| XM_017007833.1 | XP_016863322.1 | |||
| AB209181.1 | BAD92418.1 | |||
| AL834498.1 | CAD39156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830111 | Chr.4:149946205 - 150114404 on Build GRCh38 | Gain |
|
| esv3894052 | Chr.4:149995950 - 150249235 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79352672] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map