Assay Details
Target Gene Details
Entrez Gene ID: | 94120 |
Gene Name: | synaptotagmin like 3 |
Gene Aliases: |
SLP3 |
Location: |
Chr.6:158644881-158766298 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYTL3 | XM_005267215.4 | XP_005267272.1 | ||
| XM_006715605.3 | XP_006715668.1 | |||
| XM_006715606.3 | XP_006715669.1 | |||
| XM_006715611.3 | XP_006715674.1 | |||
| XM_011536254.2 | XP_011534556.1 | |||
| XM_017011495.1 | XP_016866984.1 | |||
| XM_017011496.1 | XP_016866985.1 | |||
| XM_017011498.1 | XP_016866987.1 | |||
| XM_017011499.1 | XP_016866988.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73797068] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map