Assay Details
Target Gene Details
Entrez Gene ID: | 101929122 |
Gene Name: | uncharacterized LOC101929122 |
Gene Aliases: |
- |
Location: |
Chr.6:159165899-159170007 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929122 | NR_121668.1 | 2 | 262 | |
| BX089650.1 | 2 | 262 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763604 | Chr.6:159088341 - 159521923 on Build GRCh38 | Gain |
|
| nsv1019191 | Chr.6:159134068 - 159622215 on Build GRCh38 | Gain |
|
| nsv830850 | Chr.6:159163229 - 159357812 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115313288] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map