Assay Details
Target Gene Details
Entrez Gene ID: | 135112 |
Gene Name: | nuclear receptor coactivator 7 |
Gene Aliases: |
ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3 |
Location: |
Chr.6:125780948-125932030 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Gene Details
Entrez Gene ID: | 104355145 |
Gene Name: | NCOA7 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:125797861-125818858 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCOA7-AS1 | NR_126386.1 | |||
| AW444508.1 |
More Information
Set Membership: |
|

Genomic Map