Assay Details
Target Gene Details
Entrez Gene ID: | 696 |
Gene Name: | butyrophilin subfamily 1 member A1 |
Gene Aliases: |
BT, BTN, BTN1 |
Location: |
Chr.6:26499290-26510425 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTN1A1 | NM_001732.2 | NP_001723.2 | ||
| XM_005249340.3 | XP_005249397.2 | |||
| BC096312.3 | AAH96312.1 | |||
| BC096313.1 | AAH96313.1 | |||
| BC096314.1 | AAH96314.1 | |||
| BC096315.1 | AAH96315.1 | |||
| U39576.1 | AAC50489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023453 | Chr.6:25853602 - 26528022 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map