Assay Details
Target Gene Details
Entrez Gene ID: | 2914 |
Gene Name: | glutamate metabotropic receptor 4 |
Gene Aliases: |
GPRC1D, MGLUR4, mGlu4 |
Location: |
Chr.6:34018643-34155622 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM4 | NM_000841.3 | NP_000832.1 | ||
| NM_001256809.2 | NP_001243738.1 | |||
| NM_001256811.2 | NP_001243740.1 | |||
| NM_001256812.2 | NP_001243741.1 | |||
| NM_001256813.2 | NP_001243742.1 | |||
| NM_001282847.1 | NP_001269776.1 | |||
| XM_017010790.1 | XP_016866279.1 | |||
| XM_017010791.1 | XP_016866280.1 | |||
| XM_017010792.1 | XP_016866281.1 | |||
| XM_017010793.1 | XP_016866282.1 | |||
| AA993227.1 | ||||
| AK122836.1 | ||||
| AK122982.1 | ||||
| AK126746.1 | ||||
| AK131536.1 | BAD18673.1 | |||
| AK289534.1 | ||||
| AK289563.1 | ||||
| AK293913.1 | ||||
| AK293949.1 | ||||
| AK312300.1 | ||||
| BC130526.1 | ||||
| BC130528.1 | ||||
| BC144057.1 | ||||
| EU432124.1 | ||||
| U92457.1 | AAB51762.1 | |||
| X80818.1 | CAA56784.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608630 | Chr.6:33439052 - 34076935 on Build GRCh38 | Gain |
|
| nsv602861 | Chr.6:33985869 - 34083140 on Build GRCh38 | Loss |
|
| nsv602843 | Chr.6:33964665 - 34227234 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79933334] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map