Assay Details
Target Gene Details
Entrez Gene ID: | 7430 |
Gene Name: | ezrin |
Gene Aliases: |
CVIL, CVL, HEL-S-105, VIL2 |
Location: |
Chr.6:158765741-158819424 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EZR | NM_001111077.1 | NP_001104547.1 | ||
| AK296738.1 | ||||
| AK312597.1 | ||||
| AL162086.1 | CAB82418.1 | |||
| DA754543.1 |
Target Gene Details
Entrez Gene ID: | 101409257 |
Gene Name: | EZR antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:158817979-158822239 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EZR-AS1 | NR_102425.1 | |||
| AI831881.1 | ||||
| BE552153.1 | ||||
| BX119561.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1112910 | Chr.6:158818368 - 158819068 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map