Assay Details
Target Gene Details
Entrez Gene ID: | 83741 |
Gene Name: | transcription factor AP-2 delta |
Gene Aliases: |
TFAP2BL1 |
Location: |
Chr.6:50713544-50773033 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFAP2D | NM_172238.3 | NP_758438.2 | ||
| AY028376.1 | AAK16926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608967 | Chr.6:50730716 - 50732072 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map