Assay Details
Target Gene Details
Entrez Gene ID: | 9444 |
Gene Name: | QKI, KH domain containing RNA binding |
Gene Aliases: |
Hqk, QK, QK1, QK3, hqkI |
Location: |
Chr.6:163414486-163578596 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QKI | NM_001301085.1 | NP_001288014.1 | ||
| NM_006775.2 | NP_006766.1 | |||
| NM_206853.2 | NP_996735.1 | |||
| NM_206854.2 | NP_996736.1 | |||
| NM_206855.2 | NP_996737.1 | |||
| XM_011536258.2 | XP_011534560.1 | |||
| XM_011536259.2 | XP_011534561.1 | |||
| XM_011536260.2 | XP_011534562.1 | |||
| XM_011536261.1 | XP_011534563.1 | |||
| XM_017011502.1 | XP_016866991.1 | |||
| XM_017011503.1 | XP_016866992.1 | |||
| XM_017011504.1 | XP_016866993.1 | |||
| AB067798.1 | BAB69496.1 | |||
| AB067799.1 | BAB69497.1 | |||
| AB067800.1 | BAB69498.1 | |||
| AB067801.1 | BAB69499.1 | |||
| AF142422.1 | AAF63417.1 | |||
| AF272349.1 | AAL36980.1 | |||
| AK027309.1 | BAB55032.1 | |||
| AY780788.1 | AAV98358.1 | |||
| BC019917.2 | AAH19917.1 | |||
| DQ323998.1 | ABC88600.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv949886 | Chr.6:163416369 - 163458368 on Build GRCh38 | Duplication |
|
| nsv523664 | Chr.6:163437205 - 163486853 on Build GRCh38 | Gain |
|
| nsv1019551 | Chr.6:163421326 - 163544184 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map