Assay Details
Target Gene Details
Entrez Gene ID: | 619208 |
Gene Name: | family with sequence similarity 229 member B |
Gene Aliases: |
C6orf225 |
Location: |
Chr.6:112087471-112102790 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM229B | NM_001033564.2 | NP_001028736.1 | ||
| XM_017011174.1 | XP_016866663.1 | |||
| XM_017011175.1 | XP_016866664.1 | |||
| AL049461.1 | ||||
| BC044912.2 | AAH44912.1 | |||
| BF110308.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73541435] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map