Assay Details
Target Gene Details
Entrez Gene ID: | 101409257 |
Gene Name: | EZR antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:158817979-158822239 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EZR-AS1 | NR_102425.1 | |||
| AI831881.1 | ||||
| BE552153.1 | ||||
| BX119561.1 |

Genomic Map