Assay Details
Target Gene Details
Entrez Gene ID: | 202 |
Gene Name: | absent in melanoma 1 |
Gene Aliases: |
CRYBG1, ST4 |
Location: |
Chr.6:106360808-106570449 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIM1 | XM_005266839.3 | 1 | 187 | XP_005266896.1 |
| AK125137.1 | 1 | 187 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161442 | Chr.6:106196410 - 106519212 on Build GRCh38 | Duplication |
|
| esv3610266 | Chr.6:106112750 - 106457586 on Build GRCh38 | Gain |
|
| esv3610272 | Chr.6:106259255 - 106458865 on Build GRCh38 | Gain |
|
| nsv1031707 | Chr.6:106355450 - 106448775 on Build GRCh38 | Loss |
|
| dgv6134n100 | Chr.6:106081902 - 106464172 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111350805] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map