Assay Details
Target Gene Details
Entrez Gene ID: | 135112 |
Gene Name: | nuclear receptor coactivator 7 |
Gene Aliases: |
ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3 |
Location: |
Chr.6:125780948-125932030 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Gene Details
Entrez Gene ID: | 104355145 |
Gene Name: | NCOA7 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:125797861-125818858 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCOA7-AS1 | NR_126386.1 | |||
| AW444508.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950505 | Chr.6:125772255 - 125803354 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145350186] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map