Assay Details
Target Gene Details
Entrez Gene ID: | 10667 |
Gene Name: | phenylalanyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
COXPD14, FARS1, HSPC320, PheRS |
Location: |
Chr.6:5261001-5771592 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FARS2 | NM_001318872.1 | NP_001305801.1 | ||
| NM_006567.4 | NP_006558.1 | |||
| XM_005248812.3 | XP_005248869.1 | |||
| XM_011514247.2 | XP_011512549.1 | |||
| XM_011514248.2 | XP_011512550.1 | |||
| XM_011514249.1 | XP_011512551.1 | |||
| XM_017010186.1 | XP_016865675.1 | |||
| XM_017010187.1 | XP_016865676.1 | |||
| AF097441.1 | AAC83802.1 | |||
| AF161438.1 | AAF28998.1 | |||
| AI338405.1 | ||||
| AK223423.1 | BAD97143.1 | |||
| AK312454.1 | ||||
| BC020239.1 | AAH20239.1 | |||
| BC021112.1 | AAH21112.1 | |||
| CA432122.1 | ||||
| CN292346.1 | ||||
| CR542279.1 | CAG47075.1 |
Target Gene Details
Entrez Gene ID: | 101927972 |
Gene Name: | uncharacterized LOC101927972 |
Gene Aliases: |
- |
Location: |
Chr.6:5452468-5458075 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927972 | NR_125846.1 | |||
| NR_125847.1 | ||||
| NR_125848.1 | ||||
| AI335178.1 | ||||
| CB046455.1 | ||||
| DB135850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017506 | Chr.6:5443305 - 5550892 on Build GRCh38 | Gain |
|
| esv3607971 | Chr.6:5430113 - 5461212 on Build GRCh38 | Gain |
|
| esv3607968 | Chr.6:5335705 - 5459591 on Build GRCh38 | Loss |
|
| nsv600886 | Chr.6:5426032 - 5511381 on Build GRCh38 | Loss |
|
| nsv1030744 | Chr.6:5327779 - 5457989 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map