Assay Details
Target Gene Details
Entrez Gene ID: | 5429 |
Gene Name: | DNA polymerase eta |
Gene Aliases: |
RAD30, RAD30A, XP-V, XPV |
Location: |
Chr.6:43576141-43620523 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLH | NM_001291969.1 | NP_001278898.1 | ||
| NM_001291970.1 | NP_001278899.1 | |||
| NM_006502.2 | NP_006493.1 | |||
| XM_005249186.3 | XP_005249243.1 | |||
| XM_011514698.2 | XP_011513000.1 | |||
| AB024313.1 | BAA81666.1 | |||
| AF158185.1 | AAD43810.1 | |||
| AJ420484.1 | ||||
| AK023893.1 | ||||
| AK122807.1 | ||||
| AK294430.1 | ||||
| AK307660.1 | ||||
| AK316151.1 | ||||
| BC015742.1 | AAH15742.1 |
More Information
Set Membership: |
|

Genomic Map