Assay Details
Target Gene Details
Entrez Gene ID: | 8899 |
Gene Name: | pre-mRNA processing factor 4B |
Gene Aliases: |
PR4H, PRP4, PRP4H, PRP4K, dJ1013A10.1 |
Location: |
Chr.6:4019402-4064983 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034223 | Chr.6:4030474 - 4060113 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map