Assay Details
Target Gene Details
Entrez Gene ID: | 57037 |
Gene Name: | ankyrin repeat and MYND domain containing 2 |
Gene Aliases: |
ZMYND20 |
Location: |
Chr.7:16599776-16645817 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKMY2 | NM_020319.2 | NP_064715.1 | ||
| XM_011515458.1 | XP_011513760.1 | |||
| AK001389.1 | ||||
| AK001740.1 | ||||
| AK023704.1 | ||||
| AL050390.1 | CAH56419.1 | |||
| BC015453.1 | AAH15453.1 | |||
| BC035353.1 | AAH35353.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522819 | Chr.7:16599990 - 16615762 on Build GRCh38 | Loss |
|
| dgv6289n100 | Chr.7:16230707 - 17452994 on Build GRCh38 | Gain |
|
| nsv1028125 | Chr.7:16489535 - 16807601 on Build GRCh38 | Gain |
|
| nsv830912 | Chr.7:16583571 - 16752349 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map