Assay Details
Target Gene Details
Entrez Gene ID: | 23480 |
Gene Name: | Sec61 translocon gamma subunit |
Gene Aliases: |
SSS1 |
Location: |
Chr.7:54752247-54759246 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC61G | NM_001012456.1 | NP_001012474.1 | ||
| NM_014302.3 | NP_055117.1 | |||
| AF054184.1 | AAC99401.1 | |||
| AF086539.1 | ||||
| AK308617.1 | ||||
| AK311845.1 | ||||
| BC009480.2 | AAH09480.1 | |||
| CD385140.1 | ||||
| CR456979.1 | CAG33260.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761332 | Chr.7:54574291 - 55097556 on Build GRCh38 | Gain |
|
| nsv607029 | Chr.7:54579095 - 55086688 on Build GRCh38 | Gain |
|
| esv2734425 | Chr.7:53868335 - 55917254 on Build GRCh38 | Deletion |
|
| nsv1030676 | Chr.7:54583600 - 55151489 on Build GRCh38 | Gain |
|
| nsv1024361 | Chr.7:53993970 - 54776794 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map