Assay Details
Target Gene Details
Entrez Gene ID: | 1595 |
Gene Name: | cytochrome P450 family 51 subfamily A member 1 |
Gene Aliases: |
CP51, CYP51, CYPL1, LDM, P450-14DM, P450L1 |
Location: |
Chr.7:92112149-92134745 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP51A1 | NM_000786.3 | NP_000777.1 | ||
| NM_001146152.1 | NP_001139624.1 | |||
| AK295932.1 | ||||
| AK314205.1 | ||||
| BC018429.1 | ||||
| BC032322.1 | AAH32322.1 | |||
| D55653.1 | BAA09512.1 | |||
| DC376300.1 | ||||
| U23942.1 | AAB39951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5839 | Chr.7:92123796 - 92150740 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74647299] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map