Assay Details
Target Gene Details
Entrez Gene ID: | 1780 |
Gene Name: | dynein cytoplasmic 1 intermediate chain 1 |
Gene Aliases: |
DNCI1, DNCIC1 |
Location: |
Chr.7:95772506-96110322 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DYNC1I1 | NM_001135556.1 | NP_001129028.1 | ||
| NM_001135557.1 | NP_001129029.1 | |||
| NM_001278421.1 | NP_001265350.1 | |||
| NM_001278422.1 | NP_001265351.1 | |||
| NM_004411.4 | NP_004402.1 | |||
| XM_011515863.2 | XP_011514165.1 | |||
| XM_017011804.1 | 1 | 232 | XP_016867293.1 | |
| XM_017011806.1 | XP_016867295.1 | |||
| XM_017011807.1 | XP_016867296.1 | |||
| AF063228.1 | AAC33443.1 | |||
| AF123074.1 | AAD26852.1 | |||
| AK091339.1 | BAC03639.1 | |||
| AK297427.1 | ||||
| AK313148.1 | ||||
| BC022540.1 | AAH22540.1 | |||
| BC032945.2 | AAH32945.1 | |||
| BC037854.1 | AAH37854.1 | |||
| BC053542.1 | AAH53542.1 | |||
| DA491512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
|
| nsv1026003 | Chr.7:95733543 - 96100442 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map